N-(4-Methyl-3-nitrophenyl)-4-(trifluoromethyl)benzothioamide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD269339-250MGEA 1520 DKK
BLDPBD269339-250MG BLDPBD269339-100MG
N-(4-Methyl-3-nitrophenyl)-4-(trifluoromethyl)benzothioamide 95%
N-(4-Methyl-3-nitrophenyl)-4-(trifluoromethyl)benzothioamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₅H₁₁F₃N₂O₂S
Molekylvægt: 340,32 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD27952654
CAS-nummer: 1257095-63-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR