2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)benzo[d]oxazole 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD269257-100MGEA 272 DKK
BLDPBD269257-100MG BLDPBD269257-1G BLDPBD269257-250MG
2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)benzo[d]oxazole 95%
2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)benzo[d]oxazole

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₉H₂₀BNO₃
Molekylvægt: 321,17 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 439090-73-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR