4-Fluoro-2-[(methylamino)methyl]phenol 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD268701-5GEA 3530 DKK
BLDPBD268701-5G BLDPBD268701-250MG BLDPBD268701-1G
4-Fluoro-2-[(methylamino)methyl]phenol 95%
4-Fluoro-2-[(methylamino)methyl]phenol

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₈H₁₀FNO
Molekylvægt: 155,16 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD21641325
CAS-nummer: 1363166-05-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR