10,11-Dihydrocarbamazepine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD268597-1GEA 2920 DKK
BLDPBD268597-1G BLDPBD268597-100MG BLDPBD268597-250MG
10,11-Dihydrocarbamazepine 98%
10,11-Dihydrocarbamazepine

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₅H₁₄N₂O
Molekylvægt: 238,29 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005072
CAS-nummer: 3564-73-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR