1-Bromo-2,5-dimethyl-4-nitrobenzene 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD268477-1GEA 473 DKK
BLDPBD268477-1G BLDPBD268477-25G BLDPBD268477-10G BLDPBD268477-250MG BLDPBD268477-5G
1-Bromo-2,5-dimethyl-4-nitrobenzene 98%
1-Bromo-2,5-dimethyl-4-nitrobenzene

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₈BrNO₂
Molekylvægt: 230,059 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01463298
CAS-nummer: 15540-81-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR