5-Fluoro-4-methyl-2-nitrophenol 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD268439-5GEA 6280 DKK
BLDPBD268439-5G BLDPBD268439-250MG BLDPBD268439-1G
5-Fluoro-4-methyl-2-nitrophenol 98%
5-Fluoro-4-methyl-2-nitrophenol

Storage condition: Sealed in dry, 2-8°C

Formel: C₇H₆FNO₃
Molekylvægt: 171,126 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD16658619
CAS-nummer: 83341-28-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR