2-Fluoro-4-methyl-6-nitrophenol 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD268438-100MGEA 442 DKK
BLDPBD268438-100MG
2-Fluoro-4-methyl-6-nitrophenol 95%
2-Fluoro-4-methyl-6-nitrophenol

Storage condition: Sealed in dry, 2-8°C

Formel: C₇H₆FNO₃
Molekylvægt: 171,12 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD16658596
CAS-nummer: 2070-43-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR