(E)-3-(Pyridin-3-yl)prop-2-en-1-ol 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD268422-250MGEA 528 DKK
BLDPBD268422-250MG BLDPBD268422-100MG
(E)-3-(Pyridin-3-yl)prop-2-en-1-ol 95%
(E)-3-(Pyridin-3-yl)prop-2-en-1-ol

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₉NO
Molekylvægt: 135,16 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03425627
CAS-nummer: 120277-39-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR