Methyl 4-methoxy-3,5-dimethylpicolinate 95+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD268188-100MGEA 907 DKK
BLDPBD268188-100MG BLDPBD268188-1G BLDPBD268188-250MG
Methyl 4-methoxy-3,5-dimethylpicolinate 95+%
Methyl 4-methoxy-3,5-dimethylpicolinate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₀H₁₃NO₃
Molekylvægt: 195,21 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18258414
CAS-nummer: 187222-18-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR