Methyl 3-(2-cyanopropan-2-yl)benzoate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD268113-1GEA 1000 DKK
BLDPBD268113-1G BLDPBD268113-250MG BLDPBD268113-100MG
Methyl 3-(2-cyanopropan-2-yl)benzoate 95%
Methyl 3-(2-cyanopropan-2-yl)benzoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₃NO₂
Molekylvægt: 203,23 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD19694850
CAS-nummer: 872091-85-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR