Benzyl 2-oxopyrrolidine-1-carboxylate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD268035-1GEA 714 DKK
BLDPBD268035-1G BLDPBD268035-250MG
Benzyl 2-oxopyrrolidine-1-carboxylate 95%
Benzyl 2-oxopyrrolidine-1-carboxylate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₂H₁₃NO₃
Molekylvægt: 219,23 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12827413
CAS-nummer: 14468-80-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR