2-Amino-6-methylisonicotinic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD267839-250MGEA 1780 DKK
BLDPBD267839-250MG BLDPBD267839-100MG BLDPBD267839-1G
2-Amino-6-methylisonicotinic acid 95%
2-Amino-6-methylisonicotinic acid

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₇H₈N₂O₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD13188664
CAS-nummer: 65169-64-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR