Naphthalene-1-sulphonamide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD267812-100MGEA 1190 DKK
BLDPBD267812-100MG
Naphthalene-1-sulphonamide 95%
Naphthalene-1-sulphonamide

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₀H₉NO₂S
Molekylvægt: 207,24 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00985933
CAS-nummer: 606-25-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR