Methyl 3-formyl-2-methoxybenzoate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD267722-250MGEA 1220 DKK
BLDPBD267722-250MG BLDPBD267722-100MG
Methyl 3-formyl-2-methoxybenzoate 95%
Methyl 3-formyl-2-methoxybenzoate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₁₀O₄
Molekylvægt: 194,18 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD24387418
CAS-nummer: 186312-96-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR