3-Hydroxy-1H-pyrazole-4-carboxylic acid ethyl ester 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD267715-250MGEA 117 DKK
BLDPBD267715-250MG BLDPBD267715-5G BLDPBD267715-25G BLDPBD267715-100MG BLDPBD267715-10G BLDPBD267715-1G
3-Hydroxy-1H-pyrazole-4-carboxylic acid ethyl ester 98%
3-Hydroxy-1H-pyrazole-4-carboxylic acid ethyl ester

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₈N₂O₃
Molekylvægt: 156,14 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12131138
CAS-nummer: 7251-53-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR