5'-Phenyl-[1,1':3',1''-terphenyl]-4-amine 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD267665-250MGEA 140 DKK
BLDPBD267665-250MG
5'-Phenyl-[1,1':3',1''-terphenyl]-4-amine 97%
5'-Phenyl-[1,1':3',1''-terphenyl]-4-amine
Formel: C₂₄H₁₉N
Molekylvægt: 321,42 g/mol
MDL Number: MFCD22581309
CAS-nummer: 343239-58-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR