Methyl 3-chloro-2-methoxybenzoate 95+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD267464-5GEA 1600 DKK
BLDPBD267464-5G BLDPBD267464-250MG BLDPBD267464-1G
Methyl 3-chloro-2-methoxybenzoate 95+%
Methyl 3-chloro-2-methoxybenzoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₉ClO₃
Molekylvægt: 200,61 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12547158
CAS-nummer: 92992-36-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR