3-Bromo-2-methoxybenzamide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD267463-100MGEA 125 DKK
BLDPBD267463-100MG BLDPBD267463-250MG BLDPBD267463-5G BLDPBD267463-1G
3-Bromo-2-methoxybenzamide 95%
3-Bromo-2-methoxybenzamide

Storage condition: Sealed in dry, 2-8°C

Formel: C₈H₈BrNO₂
Molekylvægt: 230,06 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18391870
CAS-nummer: 791136-88-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR