Methyl 4-(bromomethyl)-3-nitrobenzoate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD267320-250MGEA 566 DKK
BLDPBD267320-250MG BLDPBD267320-1G BLDPBD267320-5G
Methyl 4-(bromomethyl)-3-nitrobenzoate 95%
Methyl 4-(bromomethyl)-3-nitrobenzoate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₈BrNO₄
Molekylvægt: 274,06 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD12031824
CAS-nummer: 88089-94-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR