6-Bromo-4-methyl-1,2,3,4-tetrahydroquinoline 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD267313-1GEA 861 DKK
BLDPBD267313-1G BLDPBD267313-100MG BLDPBD267313-10G BLDPBD267313-25G BLDPBD267313-5G BLDPBD267313-250MG
6-Bromo-4-methyl-1,2,3,4-tetrahydroquinoline 97%
6-Bromo-4-methyl-1,2,3,4-tetrahydroquinoline

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₀H₁₂BrN
Molekylvægt: 226,11 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD26626379
CAS-nummer: 946837-99-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR