Methyl 4-Bromo-3-chlorobenzoate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD266883-1GEA 117 DKK
BLDPBD266883-1G BLDPBD266883-5G BLDPBD266883-100G BLDPBD266883-25G
Methyl 4-Bromo-3-chlorobenzoate 98%
Methyl 4-Bromo-3-chlorobenzoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆BrClO₂
Molekylvægt: 249,49 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11045619
CAS-nummer: 117738-74-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR