Ethyl 4-hydroxy-6-oxo-6,7-dihydrothieno[2,3-b]pyridine-5-carboxylate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD266858-250MGEA 1030 DKK
BLDPBD266858-250MG BLDPBD266858-100MG BLDPBD266858-1G
Ethyl 4-hydroxy-6-oxo-6,7-dihydrothieno[2,3-b]pyridine-5-carboxylate 95%
Ethyl 4-hydroxy-6-oxo-6,7-dihydrothieno[2,3-b]pyridine-5-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₀H₉NO₄S
Molekylvægt: 239,24 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22741552
CAS-nummer: 99429-68-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR