2,5-Difluoro-3-nitrobenzoic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD266741-100MGEA 528 DKK
BLDPBD266741-100MG BLDPBD266741-1G BLDPBD266741-250MG
2,5-Difluoro-3-nitrobenzoic acid 98%
2,5-Difluoro-3-nitrobenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₃F₂NO₄
Molekylvægt: 203,09 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11848537
CAS-nummer: 741721-49-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR