6-Bromo-4-(trifluoromethyl)pyridin-2(1H)-one 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD266589-1GEA 6980 DKK
BLDPBD266589-1G
6-Bromo-4-(trifluoromethyl)pyridin-2(1H)-one 97%
6-Bromo-4-(trifluoromethyl)pyridin-2(1H)-one

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₃BrF₃NO
Molekylvægt: 241,99 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD27923577
CAS-nummer: 91416-08-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR