2-(Bromomethyl)-5-methylpyrimidine hydrobromide 95+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD266579-250MGEA 1520 DKK
BLDPBD266579-250MG BLDPBD266579-1G BLDPBD266579-100MG
2-(Bromomethyl)-5-methylpyrimidine hydrobromide 95+%
2-(Bromomethyl)-5-methylpyrimidine hydrobromide

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₈Br₂N₂
Molekylvægt: 267,94 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD27923571
CAS-nummer: 1956322-78-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR