1-(4-Hydroxy-3-(hydroxymethyl)phenyl)ethanone 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD266333-250MGEA 140 DKK
BLDPBD266333-250MG BLDPBD266333-1G BLDPBD266333-25G BLDPBD266333-5G
1-(4-Hydroxy-3-(hydroxymethyl)phenyl)ethanone 95%
1-(4-Hydroxy-3-(hydroxymethyl)phenyl)ethanone

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₀O₃
Molekylvægt: 166,17 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00819312
CAS-nummer: 39235-58-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR