Ethylsulphamoyl chloride 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD266317-100MGEA 388 DKK
BLDPBD266317-100MG
Ethylsulphamoyl chloride 95%
Ethylsulphamoyl chloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₂H₆ClNO₂S
Opbevaringstemperatur: Køleskab
MDL Number: MFCD12401754
CAS-nummer: 16548-07-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR