N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-S-(triphenylmethyl)-L-cysteine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD265931-1GEA 807 DKK
BLDPBD265931-1G BLDPBD265931-100MG BLDPBD265931-250MG
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-S-(triphenylmethyl)-L-cysteine 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-S-(triphenylmethyl)-L-cysteine

Storage condition: Sealed in dry, Room Temperature

Formel: C₃₈H₃₃NO₄S
Molekylvægt: 599,75 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11973909
CAS-nummer: 944797-51-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR