2-Isopropylmorpholine hydrochloride 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD265916-100MGEA 590 DKK
BLDPBD265916-100MG BLDPBD265916-250MG
2-Isopropylmorpholine hydrochloride 97%
2-Isopropylmorpholine hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₁₆ClNO
Molekylvægt: 165,66 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD24369144
CAS-nummer: 1432678-91-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR