{3-[(Dimethylamino)methyl]phenyl}boronic acid hydrochloride 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD265577-250MGEA 187 DKK
BLDPBD265577-250MG
{3-[(Dimethylamino)methyl]phenyl}boronic acid hydrochloride 97%
{3-[(Dimethylamino)methyl]phenyl}boronic acid hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₁₄BNO₂·ClH
Molekylvægt: 215,485 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD20731144
CAS-nummer: 1485417-01-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR