3-Cyclohexylisoxazol-5-amine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD265408-250MGEA 1410 DKK
BLDPBD265408-250MG BLDPBD265408-1G BLDPBD265408-100MG
3-Cyclohexylisoxazol-5-amine 95%
3-Cyclohexylisoxazol-5-amine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₉H₁₄N₂O
Molekylvægt: 166,22 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD10693350
CAS-nummer: 500766-46-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR