6-(Trifluoromethoxy)indoline 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD265401-5GEA 5420 DKK
BLDPBD265401-5G BLDPBD265401-25G BLDPBD265401-1G BLDPBD265401-100MG BLDPBD265401-10G BLDPBD265401-250MG
6-(Trifluoromethoxy)indoline 98%
6-(Trifluoromethoxy)indoline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₈F₃NO
Molekylvægt: 203,16 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09954774
CAS-nummer: 959235-95-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR