2,3-Difluoro-5-nitrobenzoic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD264715-250MGEA 1250 DKK
BLDPBD264715-250MG BLDPBD264715-1G BLDPBD264715-100MG
2,3-Difluoro-5-nitrobenzoic acid 95%
2,3-Difluoro-5-nitrobenzoic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₃F₂NO₄
Molekylvægt: 203,09 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09859890
CAS-nummer: 942035-31-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR