1-(6-Bromoindolin-1-yl)ethanone 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD264698-1GEA 1830 DKK
BLDPBD264698-1G BLDPBD264698-250MG BLDPBD264698-100MG
1-(6-Bromoindolin-1-yl)ethanone 95%
1-(6-Bromoindolin-1-yl)ethanone

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₀BrNO
Molekylvægt: 240,09 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD15523552
CAS-nummer: 114744-53-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR