2-Chloro-4-(1H-pyrazol-1-yl)benzenecarboxylic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD264635-250MGEA 380 DKK
BLDPBD264635-250MG BLDPBD264635-1G BLDPBD264635-100MG BLDPBD264635-5G
2-Chloro-4-(1H-pyrazol-1-yl)benzenecarboxylic acid 95%
2-Chloro-4-(1H-pyrazol-1-yl)benzenecarboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₀H₇ClN₂O₂
Molekylvægt: 222,63 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD03425733
CAS-nummer: 313674-12-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR