Morpholino(quinoxalin-6-yl)methanone 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD264562-100MGEA 528 DKK
BLDPBD264562-100MG BLDPBD264562-250MG BLDPBD264562-1G
Morpholino(quinoxalin-6-yl)methanone 97%
Morpholino(quinoxalin-6-yl)methanone

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₁₃N₃O₂
Molekylvægt: 243,261 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01444315
CAS-nummer: 312319-71-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR