3-Fluoro-4-piperidinone hydrochloride 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD264526-5GEA 1140 DKK
BLDPBD264526-5G BLDPBD264526-250MG BLDPBD264526-1G
3-Fluoro-4-piperidinone hydrochloride 98%
3-Fluoro-4-piperidinone hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₅H₉ClFNO
Molekylvægt: 153,58 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11506659
CAS-nummer: 1070896-59-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR