Methyl-3-cyanoindole-6-carboxylate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD264480-1GEA 1830 DKK
BLDPBD264480-1G BLDPBD264480-100MG BLDPBD264480-250MG
Methyl-3-cyanoindole-6-carboxylate 98%
Methyl-3-cyanoindole-6-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₁H₈N₂O₂
Molekylvægt: 200,197 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09878564
CAS-nummer: 1000576-51-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR