3,3'-Oxybis(propan-1-ol) 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD264351-1GEA 737 DKK
BLDPBD264351-1G BLDPBD264351-10G BLDPBD264351-5G BLDPBD264351-250MG
3,3'-Oxybis(propan-1-ol) 98%
3,3'-Oxybis(propan-1-ol)

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₁₄O₃
Molekylvægt: 134,17 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00051023
CAS-nummer: 2396-61-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR