tert-Butyl (2-bromo-4,5,6,7-tetrahydrobenzo[d]thiazol-6-yl)carbamate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD264062-100MGEA 776 DKK
BLDPBD264062-100MG BLDPBD264062-1G BLDPBD264062-250MG
tert-Butyl (2-bromo-4,5,6,7-tetrahydrobenzo[d]thiazol-6-yl)carbamate 95%
tert-Butyl (2-bromo-4,5,6,7-tetrahydrobenzo[d]thiazol-6-yl)carbamate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₂H₁₇BrN₂O₂S
Molekylvægt: 333,25 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD13195380
CAS-nummer: 1251009-79-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR