2-Amino-4-bromobenzyl alcohol 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD263940-1GEA 318 DKK
BLDPBD263940-1G BLDPBD263940-5G BLDPBD263940-25G BLDPBD263940-250MG BLDPBD263940-10G
2-Amino-4-bromobenzyl alcohol 97%
2-Amino-4-bromobenzyl alcohol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₈BrNO
Molekylvægt: 202,049 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07779492
CAS-nummer: 946122-05-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR