Spiro[cyclopropane-1,3'-indolin]-2'-one 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD263901-100MGEA 489 DKK
BLDPBD263901-100MG BLDPBD263901-5G BLDPBD263901-250MG BLDPBD263901-1G
Spiro[cyclopropane-1,3'-indolin]-2'-one 95%
Spiro[cyclopropane-1,3'-indolin]-2'-one

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₀H₉NO
Molekylvægt: 159,19 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11934419
CAS-nummer: 13861-75-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR