[1,2,4]Triazolo[4,3-a]pyrimidin-3(2H)-one 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD263880-1GEA 4420 DKK
BLDPBD263880-1G BLDPBD263880-250MG BLDPBD263880-100MG
[1,2,4]Triazolo[4,3-a]pyrimidin-3(2H)-one 95%
[1,2,4]Triazolo[4,3-a]pyrimidin-3(2H)-one

Storage condition: Sealed in dry, 2-8°C

Formel: C₅H₄N₄O
Molekylvægt: 136,11 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD13193348
CAS-nummer: 63206-77-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR