5-(Trifluoromethyl)-1H-pyrazol-3-amine 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD26362-250MGEA 171 DKK
BLDPBD26362-250MG BLDPBD26362-25G BLDPBD26362-5G BLDPBD26362-1G BLDPBD26362-100MG
5-(Trifluoromethyl)-1H-pyrazol-3-amine 97%
5-(Trifluoromethyl)-1H-pyrazol-3-amine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₄H₄F₃N₃
Molekylvægt: 151,09 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD06738292
CAS-nummer: 852443-61-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR