2-Amino-4-bromobenzamide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD263468-100MGEA 264 DKK
BLDPBD263468-100MG BLDPBD263468-250MG BLDPBD263468-10G BLDPBD263468-1G BLDPBD263468-5G
2-Amino-4-bromobenzamide 95%
2-Amino-4-bromobenzamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₇BrN₂O
Molekylvægt: 215,047 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12674809
CAS-nummer: 112253-70-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR