Ethyl 2,5-dibromothiophene-3-carboxylate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD263434-250MGEA 559 DKK
BLDPBD263434-250MG BLDPBD263434-100MG
Ethyl 2,5-dibromothiophene-3-carboxylate 95%
Ethyl 2,5-dibromothiophene-3-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₇H₆Br₂O₂S
Molekylvægt: 313,99 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD12032144
CAS-nummer: 289470-44-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR