5-Methylthiazole-4-carboxylic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD26332-250MGEA 140 DKK
BLDPBD26332-250MG BLDPBD26332-1G BLDPBD26332-100MG
5-Methylthiazole-4-carboxylic acid 98%
5-Methylthiazole-4-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C5H5NO2S
Molekylvægt: 143,1600 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06738813
CAS-nummer: 120237-76-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR