3-(2-Hydroxyethyl)pyrrolidin-2-one 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD263137-50MGEA 4180 DKK
BLDPBD263137-50MG BLDPBD263137-1G BLDPBD263137-100MG BLDPBD263137-250MG
3-(2-Hydroxyethyl)pyrrolidin-2-one 95%
3-(2-Hydroxyethyl)pyrrolidin-2-one

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₁₁NO₂
Molekylvægt: 129,15 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD19219407
CAS-nummer: 932-47-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR