4-((4-Amino-6-chloropyrimidin-2-yl)amino)benzonitrile 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD262831-100MGEA 884 DKK
BLDPBD262831-100MG BLDPBD262831-250MG BLDPBD262831-1G
4-((4-Amino-6-chloropyrimidin-2-yl)amino)benzonitrile 95%
4-((4-Amino-6-chloropyrimidin-2-yl)amino)benzonitrile

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₁H₈ClN₅
Molekylvægt: 245,67 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD24682776
CAS-nummer: 1398507-08-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR