2,2,4,4-Tetramethyl-1,3-cyclobutanediol (mixture of isomers) 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD262830-25GEA 528 DKK
BLDPBD262830-25G BLDPBD262830-5G
2,2,4,4-Tetramethyl-1,3-cyclobutanediol (mixture of isomers) 98%
2,2,4,4-Tetramethyl-1,3-cyclobutanediol (mixture of isomers)

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₁₆O₂
Molekylvægt: 144,21 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001329
CAS-nummer: 3010-96-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR